Learn More
Thermo Scientific™ Venetoclax, 95%
Brand: Thermo Scientific™ 468231000
92.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Additional Details : CAS Number : 1257044-40-8 Weight : 0.00010kg
Specifications
Venetoclax | |
100 mg | |
868.45 | |
Reagent |
1431612-23-5 | |
C45 H50 Cl N7 O7 S | |
Solid | |
95% |
Safety and Handling
- Venetoclax
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 4-(4-((2-(4-Chlorophenyl)-4,4-dimethylcyclohex-1-e n-1-yl)methyl)piperazin-1-yl)-N-((3-nitro-4-((tet