Learn More
Emma Safety Footwear Venus XD S3 Safety Shoes
The Venus is a low profile, easy on / easy off shoe. The smooth, easy to clean contours offer additional hygiene advantages over and above lace up shoes. It's perfect for a 'quick change' from office to factory floor or walking between one facility and another.
Brand:  Emma Safety Footwear MM79456641
 View more versions of this product
Product Code. 15929051
  • £123.00 / 1 pair
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S2-Safety shoe for wet working conditions
  • Full grain leather construction
  • GripForce™ Easy Twist
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Specifications
Venus XD S3
Leather
41