Learn More
Veolia Water Systems Veolia Water Systems™ Sterilization Tablet
Reliable and effective chlorine tablets that reduce bio film and protect the system against bacterial growth.
Brand: Veolia Water Systems CT1
Code : A0
Additional Details : Weight : 0.03420kg Transport : UN number : 3077 Chem class : 9 Pack group : III
Description
- CT1 Tablets (pack of 12) - Chlorine tablets
- Sanitizes tubing or pipe work to remove bacteria
- Reduces bio film and protect the system against bacterial growth
- Sterilization tablet
Specifications
12/Pk. |
Safety and Handling
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P270-Do not eat, drink or smoke when using this product.
Supplemental information
- MIXTURE LIST-Contains : troclosene sodium, dihydrate, boric acid