Learn More
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
13175950
|
Veolia Water Systems™
UVEGBU2009 |
UHQ6 Ultrapure Water System |
£124.00
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
||||||
13135980
|
Veolia Water Systems™
LC210-02 |
Used as a bactericide and to break down and photo-oxidize organic contaminants to polar or ionized species |
£979.00
Each |
Estimated Shipment: 05-03-2025
Log in to see stock. |
||||||
11974897
|
Veolia Water Systems™
LC118 |
Centra R 60/120, Maxima MK1/MK2, PureLab Option-Q, PureLab Ultra MK1, PureLab Ultra MK2 |
£798.00
Each |
Estimated Shipment: 05-03-2025
Log in to see stock. |
||||||
Description
- UV light is used as a bactericide and to break down and photo-oxidise organic contaminants to polar or ionized species
- The UV bactericidal action damages DNA and RNA polymerase at low doses thereby preventing replication
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Veolia Water Systems™ UV Lamp