Learn More
Emma Safety Footwear Vera Safety Shoes
An easy shoe for wipe clean hygiene. The upper is completely washable microfiber construction. Sanitized with Hydro-Tec™ Sanitized Silver. Stain, fat, grease and direct resistant.
Brand:  Emma Safety Footwear MM50654439
Additional Details : Weight : 1.33100kg
 View more versions of this product
Product Code. 15908561
  • £140.00 / 1 pair
Estimated Shipment: 05-09-2024
to see stock.
Add to basket
Description
Description
  • S2-Safety shoe for wet working conditions
  • GripForce™ Easy Twist
  • Microfiber construction
  • ESD certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver
Specifications
Specifications
EN ISO 20345:2011
White
Hydro-Tec™ Sanitized Silver
39
Elastic
Vera
Microfiber
Steel