missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Verapamil hydrochloride, 99.9%, MP Biomedicals™
Click to view available options
Quantity:
5 g
Specifications
CAS | 52-53-9 |
Color | White |
Assay Percent Range | 0.999 |
Molecular Formula | C27H38N2O4 |
Quantity | 5 g |
Synonym | verapamil, iproveratril, vasolan, dilacoran, isoptin, cordilox, calan, verapamilo, verapamilum, falicard |
InChI Key | SGTNSNPWRIOYBX-UHFFFAOYSA-N |
SMILES | CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC |
IUPAC Name | 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile |
Molecular Weight (g/mol) | 454.611 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
Product Identifier
- (±)-Verapamil Hydrochloride
- Danger
- Acute toxicity Category 3
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : verapamil hydrochloride
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'