missing translation for 'onlineSavingsMsg'
Learn More
Verapamil hydrochloride, Tocris Bioscience™ Product Code.: 18732544

Verapamil hydrochloride, Tocris Bioscience™

Product Code. 18732544
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 18732544

Brand: Tocris Bioscience 0654/1G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CaV1.x blocker

Verapamil hydrochloride is an L-type calcium channel blocker. Vasodilator and adrenergic antagonist. Verapamil induces autophagy by modulating cellular metabolism. Verapamil has been identified as targeting human host proteins that interact with SARS-CoV-2.

Specifications

CAS 152-11-4
Molecular Formula C27H39ClN2O4
Quantity 1 g
Synonym verapamil hydrochloride, verapamil hcl, manidon, calcan hydrochloride, cardibeltin, cardiabeltin, cardioprotect, veroptinstada, calaptin, caveril
InChI Key DOQPXTMNIUCOSY-UHFFFAOYSA-N
SMILES CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC.Cl
IUPAC Name 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile;hydrochloride
Molecular Weight (g/mol) 491.069
PubChem CID 62969
Formula Weight 491.07
Percent Purity 0.98
Chemical Name or Material Verapamil hydrochloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Verapamil hydrochloride, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.