Learn More
Cole-Parmer™ Rapid Surface Hygiene
Detects invisible food residue without an instrument -in only 10 to 60 seconds
Brand:  Cole-Parmer™ 14040-30
Product Code. 11740559
  • £148.00 / Pack of 50
Explore more special offers
Description
Description
  • VERIcleen is sensitive to food residues including meat, dairy, bakery, beverages, eggs, fish, etc. It is an inexpensive rapid hygiene program to the foodservice, retail and institutional industries.
  • Applications include HACCP programs, hand washing hygiene compliance, and overall sanitation monitoring.
  • Application is extremely simple, just spray to wet-dry surface and swab.
  • A “clean surface” shows no purple until after 3 minutes. A “Not clean” surface shows purple in 10 to 60 seconds.
  • Each kit includes 50 tests in a light-tight snap-top container, 1 bottle of wetting liquid with an atomizer, instruction leaflet, and desiccant packet (if desiccant inside the packet has turned from blue to pink, the strips have been stored improperly, and may be invalid).
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Rapid Surface Hygiene > Detects: Invisible Food Residue