Learn More
Gilson™ VERITY 4220 Dual Syringe Pump
The VERITY™ Syringe Pumps provide speed and reliability to small- and large-volume, repetitive liquid transfers in HPLC, SPE, and other general liquid handling methods. These pumps work with many VERITY systems and are available in single syringe, dual syringe, and dual syringe with tee configurations. A pressure sensor is available in some models.
Brand:  Gilson™ 31130003
Product Code. 16616165
  • £9510.00 / Each
Estimated Shipment: 28-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Choose the Right Pump for Your Needs
  • VERITY 4000-Series Syringe Pumps are ideal for all VERITY GX-series liquid handlers and the 223 Sample Changer.
  • VERITY 4060 and 4260 Syringe Pumps are designed for use with VERITY ASPEC™ SPE systems to automate liquid handling and extractions using positive pressure elution.
  • The VERITY 4060 Single Syringe Pump and VERITY 4260 Dual Syringe Pump when paired with an ASPEC system, can automate liquid handling procedures and solid phase extraction (SPE) using positive pressure elution.
Specifications
Specifications
100/240 V
22.6 x 17.1 x 26.9 cm
5°C
7.1 kg
40°C