missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vermiculite, Honeywell™ Fluka™
Click to view available options
Quantity:
10 kg
Packaging:
Paper/Poly Bag
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Tan |
Quantity | 10 kg |
Melting Point | 1000°C |
CAS | 1318-00-9 |
Packaging | Paper/Poly Bag |
MDL Number | MFCD00132837 |
UN Number | NONH for all modes of transport |
Chemical Name or Material | Vermiculite |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'