Learn More
3M™ Versaflo™ S-Series Hood and Head Covers Assemblies
Tucking the inner shroud into a shirt or protective coverall allows excess air to be channeled over the body, providing additional comfort. The 3M Versaflow S-Series Hood Assembly is one-size-fits-all.
Brand: 3M™ 7100079874
153.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Code : S8
Additional Details : Weight : 0.84600kg
Description
Manufactured from general purpose cost effective material, with knitted inner collar. The reusable suspension hoods can be a highly cost effective for some high disposal rate environments: just the soiled hood fabric is replaced, while the suspension and air ducting can be reused. It also has an externally-adjustable airflow control that allows wearers to modify the airflow distribution to suit personal comfort needs.
- Hood with reusable suspension providing head, face, neck and shoulder coverage
- Provides eye and face protection to EN166 - liquid splash and low energy impact (grade F) - the same classification for impact resistance as safety spectacles
- Externally adjustable airflow to suit the wearers needs
- Highly cost effective for high disposal rate environments
- General purpose, cost effective fabric, with knitted collar
- Easy Disposal - components can be separated
- Replaceable over-hoods
Specifications
Gray | |
Respirator Hood | |
3M Powered and supplied air respirator systems | |
S-855E |
Spunbound Polypropylene | |
Standard | |
Head harness and S-805E replacement fabric hood | |
1 Ea. |