missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Versaflo™ M-200 Series Faceshield
3M™ Versaflo™ Faceshields, M-200 Series feature lightweight, compact and well-balanced faceshield that can offer integrated protection from a range of respiratory, head, eye, face and hearing hazards (when used with optional hearing protection).
£291.00
Specifications
Color | Light Gray |
---|---|
Disposable | Reusable |
Filter Type | None |
Material | Polyethylene |
Product Type | Faceshield |
Product Code | Brand | Design Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Design Type | Price | Quantity & Availability | |||||
17825666
|
3M™
7100258667 |
Comfort Faceseal |
£291.00
Each |
Estimated Shipment: 09-09-2024
Log in to see stock. |
|||||
17855666
|
3M™
7100256071 |
Flame Resistant |
£291.00
Each |
Estimated Shipment: 09-09-2024
Log in to see stock. |
|||||
Description
- Get highly versatile, integrated protection from a range of respiratory, face, eye and bump risk hazards with 3M™ Versaflo™ Faceshields, M-200 Series.
- Our lightweight, compact and well-balanced faceshields feature various options including flame resistant faceseals for applications with hot particles. Designed to guard against dusts, spraying and chemical processing the headtops can be used in industries as varied as agriculture, construction, food and beverage manufacturing, medical and healthcare, paper processing, pharmaceutical manufacturing, and woodworking.
- The fully-adjustable suspension includes a textile harness for correct fit and weight distribution, and the deflector allows users to direct the airflow inside the headtop for increased control and comfort.
- The visor offers good peripheral and downward vision with good optical clarity and features a polycarbonate coating for added scratch and chemical resistance.
- They meet respiratory performance requirements (TH2 or TH3) when used with specific 3M™ Air Delivery Units and are easy to maintain with spare parts and accessories that are quick, easy and intuitive to replace.
- There are options to combine with a range of 3M™ Peltor™ hearing protection including 3M™ Peltor™ LiteCom Solutions.
Specifications
Light Gray | |
None | |
Faceshield | |
No |
Reusable | |
Polyethylene | |
Standard |