missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Versaflo™ M-200 Series Faceshield
Versaflo™ M-200 Series Faceshields are lightweight and compact, offering integrated protection against dust, spraying, and chemical processing. Supplied with various faceseal options. Feature an adjustable textile harness and airflow deflector. Can be used with 3M™ PAPR and SAR Systems.
£293.00 - £325.00
Specifications
Assigned Protection Factor (APF) | NPF: 200 (V-Series), NPF: 50 (Jupiter), NPF: 500 (TR-302E/TR-602E) |
---|---|
Certifications/Compliance | CE Approved, EN 12941 TH2/TH3, EN 14594 3B, EN 166:2001 |
Color | Light Gray |
Disposable | Reusable |
Material | Polyethylene |
Description
- Product Type: Faceshield
- Material: Polyethylene
- Color: Light Grey
- Dimensions (L x W x H): 343 mm x 343 mm x 248 mm
- Weight (Metric): 591 g
- Features: Chemical and Scratch Resistant Lenses, Large Viewing Area, Lightweight, Easy Visor Replacement System
- Protection Level: NPF: 200 (V-Series), NPF: 50 (Jupiter), NPF: 500 (TR-302E/TR-602E)
- Specifications: CE Approved, EN 12941 TH2/TH3, EN 14594 3B, EN 166:2001
- Environmental Conditions: High Mobility Required
- Hazard Type: Gas, Vapour and Particulates, Liquid Splash, Low Energy Flying Particles / Dust
- For Use with (Equipment): 3M™ PAPR and SAR Systems
- For Use with (Applications): Construction, Transportation, Automotive, Metal Production and Fabrication, Paint Spraying
Specifications
NPF: 200 (V-Series), NPF: 50 (Jupiter), NPF: 500 (TR-302E/TR-602E) | |
Light Gray | |
Polyethylene | |
Construction, Transportation, Automotive, Metal Production and Fabrication, Paint Spraying | |
Non-sterile | |
Yes | |
13.5 in. | |
13.5 in. | |
9.8 in. | |
1.3 lb. |
CE Approved, EN 12941 TH2/TH3, EN 14594 3B, EN 166:2001 | |
Reusable | |
Faceshield | |
3M PAPR and SAR Systems | |
Textile | |
343 x 343 x 248 mm | |
343 mm | |
343 mm | |
248 mm | |
591 g |