missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Versaflo™ M-300 Series Helmet
Versfalo™ Helmets, M-300 Series feature lightweight, compact and well-balanced helmets that can offer integrated protection from a range of respiratory, head, eye, face and hearing hazards, making it suitable for use on construction sites.
£476.00
Specifications
Color | Black |
---|---|
Disposable | Reusable |
Filter Type | None |
Product Type | Respirator Helmet |
Size | Standard |
Product Code | Brand | Design Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Design Type | Price | Quantity & Availability | |||||
17809271
|
3M™
7100257936 |
Flame Resistant |
£476.00
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
17835666
|
3M™
7100257937 |
Comfort Fit |
£476.00
Each |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||||
Description
- Get highly versatile, integrated protection from a range of respiratory, face, and eye hazards with 3M™ Versaflo™ Helmet M-300 Series.
- Models available include the Versaflo™ M-306 featuring a general purpose faceseal suitable for construction applications and heavy industry or the Versalo™ M-307 featuring a flame resistant faceseal, suited for applications with hot particles such as smelting, foundries and other metal working applications.
- Our helmet provides head protection approved to EN397, making it a suitable option for construction sites where respiratory protection is needed.
- The fully-adjustable suspension includes a textile harness for optimal fit and weight distribution, and the deflector allowing users to direct the airflow inside the headtop for increased control and comfort.
- The visor offers good peripheral and downward vision with good optical clarity and coated polycarbonate for added scratch and chemical resistance.
- It meets the highest respiratory performance requirements (TH3) for this type of product when used with specific 3M™ Air Delivery Units, and it’s easy to maintain with spare parts and accessories that are quick, easy and intuitive to replace.
- Can be used with a range of 3M™ Peltor™ hearing protection as well as an in-built microphone option for ease of communication.
Specifications
Black | |
None | |
Standard |
Reusable | |
Respirator Helmet |