missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Versaflo™ M-400 Series Respiratory Helmet Assembly
Used with an approved air source, M-400 Series Helmets provide respiratory (OSHA APF=1000) and limited head (ANSI Z89.1-2003 Type 1 Class G) and eye (ANSI Z87.1-2010) protection.
£697.00
Specifications
Assigned Protection Factor (APF) | 1,000 |
---|---|
Certifications/Compliance | CE Approved, EN 12941 TH3, EN 14594 3B, EN 166:2001, EN 397 |
Color | Black |
Product Type | Respirator Helmet |
For Use With (Application) | Demolition, Metalworking, Metal Processing, Smelting, Foundries (Grinding, Polishing), Woodworking, Construction |
Product Code | Brand | Model | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Includes | Price | Quantity & Availability | ||||
11793916
|
3M™
7000104038 |
M-407 | Flame-Resistant Shroud | N/A | |||||
Description
Features:
- Chemical and Scratch Resistant Lenses
- Large Viewing Area
- Lightweight, Easy Visor Replacement System
Environmental Conditions:
- Cold Working Environment
- High Mobility Required
- Safety Helmet Required
Hazard Type:
- Gas, Vapour and Particulates
- Liquid Splash
- Low Energy Flying Particles/Dust
- Falling Objects
- Sparks and Hot Particles
Includes:
- The M-405 assembly includes a M-925 Visor, M-444 Inner Collar, and M-445 Outer Shroud.
- The M-925 visor is made of polycarbonate.
- The M-445 shroud is appropriate for general purpose use.
- The M-407 Assembly includes a M-927 Premium Visor, M-444 Inner Collar, and M-447 Flame Resistant Outer Shroud.
- The M-927 visor is made of coated polycarbonate.
- The M-447 shroud is made of Nomex flame resistant fibers.
- 3M Catalog# M-405 , 3M Catalog# M-407
Specifications
1,000 | |
Black | |
Demolition, Metalworking, Metal Processing, Smelting, Foundries (Grinding, Polishing), Woodworking, Construction | |
Non-sterile | |
No | |
13.6 in. | |
13.6 in. | |
9.6 in. | |
2.4 lb. |
CE Approved, EN 12941 TH3, EN 14594 3B, EN 166:2001, EN 397 | |
Respirator Helmet | |
3M Air Delivery Units | |
Textile | |
346 x 346 x 254 mm | |
346 mm | |
346 mm | |
254 mm | |
1066 g |