Learn More
3M™ Versaflo™ M-Series Faceshields and Helmets
3M™ Versaflo™ M-Series Faceshields and Helmets can help protect your workers from a variety of respiratory and impact hazards while maintaining comfort. They are are lightweight, compact and easy to maintain.
Brand:  3M™ 7100257936
343.79 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 17809271
  • £520.00 / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • M-Series are specifically designed to work together with your 3M™ Versaflo™ Powered Air Respirator and 3M™ Supplied Air Respirator Systems.
  • Deflector allows users to direct the airflow inside the headtop to increase comfort.
  • Fully-adjustable suspension for customized fit and weight distribution.
  • Accessory options availalble for protection to help handle the environments you encounter.
  • Visor combines good peripheral and downward vision.
  • Specific headtops can be used with a range of 3M™ Peltor™ hearing protection as well as an attachable microphone for ease of communication.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black
Reusable
Flame Resistant Polyester
Standard
Flame Resistant Faceseal
Flame Resistant
None
Respirator Helmet
Yes
Polycarbonate Visor