Learn More
3M™ Versaflo™ Powered Air Respirator Particulate Filter
Versaflo™ Powered Air Respirator Filters, TR-300+ Series provide protection against dusts, mists and other particulate hazards when use with 3M™ Versaflo Powered Air Respirator, TR-300 Series and a 3M™ approved headtop.
Brand: 3M™ 7100266123
126.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
- Get protection against dust, mist and fumes, and nuisance acid gases or organic vapours with 3M™ Versaflo™ Powered Air Respirator Filters, TR-300+ Series.
- Our filters are designed to be used with 3M™ Versaflo™ Powered Air Turbo TR-300+ and are CE approved.
- Certain filters provide protection against nuisance level Organic Vapours or Acid Gases (concentrations below the occupational exposure limit).
- Filters are easy to connect to the turbo unit and the 3M™ Versaflo™ Powered Air Respirator, TR-302E features an audible and visual alarm to let you know when to replace your filter.
Specifications
COMBINATION | |
Various | |
Standard | |
Versaflo PAPR | |
2 |
Gray | |
Filter and Cartridge | |
High Efficiency | |
TR-300+ |