Learn More
3M™ Versaflo™ Powered Air Respirator System Easy Clean Ready Kit, TR-600E ECK
3M™ Versaflo™ TR-600 Powered Air Respirators are versatile and easy-to-use respiratory protection against certain gases and vapours and as well as particulates.
Brand: 3M™ 7100258808
1334.37 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Respiratory protection against dusts, mists, fumes and other particulate hazards, and a variety of gas and vapour hazards, including organic vapou, acid gas, inorganic gases, ammonia and mercury vapours
- Three user selectable airflows with loose-fitting and tight-fitting options, enable the user to control flow settings to allow the user to find the setting they find most comfortable
- Powered Air Respirators automatically adjust airflow to compensate for filter loading to deliver consistent filtered air during use
- Audible, visual, and vibratory alerts for low airflow and low battery charge-level to keep you prepared and informed
- High capacity battery option available for longer run time and reduced downtime
- Available in convienient, easy to buy assemblies and kits
Specifications
Black, White | |
Standard | |
Belt-Mounted |
PAPR Healthcare Systems and Kits | |
TR-6712E Filter |
Product Content Correction
Product Title
3M™ Versaflo™ Powered Air Respirator System Easy Clean Ready Kit, TR-600E ECK > Product Type: PAPR System Kit, Color: Black, Model: TR-600E ECK
Spot an opportunity for improvement?Share a Content Correction