Learn More
3M™ Versaflo™ S-Series Headcovers and Hoods
3M™ Versaflo™ S-Series Headcovers and Hoods offer a variety of options for faceseals, collars, shrouds, and suspensions, allowing you to choose the respiratory protection that provides the most value and comfort for your team.
Brand: 3M™ 7100079873
702.40 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- S-Series respiratory headcovers and hoods are specifically designed to work together with your Versaflo™ Powered Air Respirator and 3M™ Supplied Air Respirator Systems.
- Provides head, face, neck and shoulder coverage from dust and paint spray.
- Provides a good field of view.
- Helps protect eyes and face from liquid splash and low energy flying particles and dust.
- Versatile accessories available that can help you adapt the hood to your particular environment.
Specifications
1000 | |
Premium Replacement Hood with Sealed Seams and Inner Collar, S-805-5 | |
Zytron 200 | |
Assembly, facility maintenance, machine operation, painting, sanding | |
Hood with neck collar only | |
5/Cs. | |
Acetate |
NIOSH | |
Gray | |
Hood with Neck Collar | |
GVP, Breathe-Easy™, V Series, T-300, T-600, T-800 | |
S-805-5 | |
Premium Resusable Suspension |