Learn More
3M™ Versaflo™ TR-600 PAPR Accessory, Filter
3M Versaflo™ TR-600 PAPR Accessory, Filter is used with 3M™ Versaflo™ TR-600 series air purifying device.
Brand: 3M™ 7100020999
439.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.99900kg
Description
- Simple to install
- For use with 3M Versaflo Powered Air TR-600 series
- Recommends use of TR-6100FC filter cover to help protect filter from physical damage
Specifications
ABE2K1 HgP | |
3M Versaflo Powered Air Turbo TR-600 Series | |
TR-6580E |