Learn More
3M™ Versaflo™ TR-600 PAPR Accessory, Powered Air Turbo Unit, TR-602E
Product Code. 15529305
Click to view available options
:
Powered Air Turbo Unit, TR-602E
Product Code. 15529305
Brand: 3M™ 7100035056
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with 3M™ Versaflo™ TR-600 series. 3M Versaflo™ TR-600 PAPR Accessory, Powered Air Turbo Unit, TR-602E includes Airflow Indicator (TR-971) and Spark Arrestor (TR-662 x2).
  • Includes an Airflow Indicator (TR-971) and Spark Arrestor (TR-662 x2)
  • Comfortable Airflow - Three user selectable flow rates to maximise comfort. Controlled airflow delivers steady flow as battery discharges or filter loads with particulates
  • Display shows battery charge status and particulate filter loading status during use
  • Charge status is also indicated by LED display on battery
  • Electronic audible, visual and vibratory alarm in case of low battery and/or low air flow
  • No user calibration of turbo required
  • Two belt options available: Easy Clean Belt - submersible belt with smooth surfaces allows easy, thorough cleaning and High Durability Belt for hot work or rugged environments which provides greater durability and spark/flame resistance
  • Approved to EN 12941 Class TH2 or TH3 dependent on headtop.
  • Inlet plugs for full submersion and cleanup, meets IP54 and IP67 ratings for use in decontamination shower and temporary submersion when used with cleaning and storage plugs
TRUSTED_SUSTAINABILITY
Specifications
Product Type Powered Air Turbo Unit
For Use With (Equipment) 3M Versaflo TR-600 series
Includes Airflow Indicator (TR-971) and Spark Arrestor (TR-662 x2)
Model TR-602E
Weight (Metric) 670 g