Learn More
3M™ Versaflo™ Intrinsically Safe Powered Air Turbo Starter Kit, TR-819E
3M™ Versaflo™ Intrinsically Safe Powered Air Turbo Starter Kit TR-819E is a handy kit approved to Zone 0 requirements and may be suitable for areas where potentially explosive atmospheres are expected and permanent within normal operations. Ready-to-use and has everything you need to get started. Approved for use with a wide range of Versaflo™ Soft Headtops S Series and Rigid Headtops M Series. ATEX certified and CE approved.
Brand: 3M™ 7100169180
1380.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 3.38500kg Transport : UN number : 3481 Chem class : 9
Description
- Convenient ready-to-use kit
- Approved for use with a wide range of Versaflo™ Soft Headtops S Series and Rigid Headtops M Series
- Includes a rechargeable standard capacity lithium Ion battery
- Headtop not included
- CE approved
Specifications
Lithium Ion | |
CE Approved | |
PAPR System for Heavy Industry | |
1/Cs. |