Learn More
Thermo Scientific™ VersaLid for VersaCool™ Refrigerated Circulating Bath, reversible hinged lid
Reversible hinged lid to be used with Thermo Scientific VersaCool Refrigerated Circulating Bath.
Brand:  Thermo Scientific™ 230000002
Additional Details : Weight : 4.00000kg
Product Code. 15489529
  • £282.24 / Each
Estimated Shipment: 29-08-2024
to see stock.
Add to basket
Description
Description
Utilize the VersaLid for optimal performance with the Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
Configurable to the user’s comfort, the VersaLid offers a tool-less setting for left or right-handed hinge movement and also for no hinge movement. It’s dripless when used in hinge mode.
  • One lid that can be left-handed hinge, right-handed hinge, or unhinged lid 
  • Drip-less lid reduces moisture and directs condensate back into bath
Specifications
Specifications
VersaLid-reversible hinged lid
Thermo Scientific VersaCool Refrigerated Circulating Bath
Cover