Learn More
Ansell™ Versatouch™ 72-286 Series Dyneema™ Fiber Medium Weight Gloves
Product Code. 12649761 Shop All Ansell Products
7
Click to view available options
Size:
6
7
8
9
10
Exceptional cut resistance, comfort and durability. Suitable for contact with all foodstuffs.
  • Washable up to 90°C
  • Offers comfort and durability
  • Knitted construction
  • Polypropylene Glass fiber Dyneema™ liner
  • Knitwrist cuff
TRUSTED_SUSTAINABILITY
Specifications
Back Material Dyneema™ Fiber
Certifications/Compliance CE Mark, Standard CATEGORY II EN 388 : 254X
Cuff Material Dyneema™ Fiber
Product Type Mediumweight Gloves
Size 7
Glove Cuff Style Knit
Liner Material Dyneema™ Fiber
Lining Dyneema™ Fiber
Quantity 72 Pairs
Resistant To Cut resistant
Product Content Correction
Product Title
Ansell™ Versatouch™ 72-286 Series Dyneema™ Fiber Medium Weight Gloves >