missing translation for 'onlineSavingsMsg'
Learn More

Ansell VersaTouch™ 92-471 Series Light Blue Nitrile Gloves

Made from innovative nitrile compound which offers good chemical and mechanical properties, comfort and dexterity.

£24.15

Specifications

Certifications/Compliance CE Mark, Standard CATEGORY III EN 374
Product Type Controlled Environment Gloves
Sterility Non-sterile
Thickness (Metric) 0.12 mm
Quantity 100 Pairs
View More Specs

Products 4
Product Code Brand dimSize Price Quantity & Availability  
Product Code Brand dimSize Price Quantity & Availability  
12687061
View Documents
Ansell
92471070
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12607071
View Documents
Ansell
92471090
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12697061
View Documents
Ansell
92471080
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12617071
View Documents
Ansell
92471100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Textured finish on fingertips
  • Improved grip
  • Contains no natural rubber
  • Suitable for contact with all fatty foods
  • Outstanding resistance to animal fats compared with PVC or natural rubber
  • Powder-free
  • No risk of contaminating food with corn starch used to powder gloves

Applications:

  • All final stages of meat or fish processing
  • Controlled-environment rooms
  • Prepared meals
  • Dairy processing
  • Catering
Specifications

Specifications

CE Mark, Standard CATEGORY III EN 374
Non-sterile
100 Pairs
92-471
Controlled Environment Gloves
0.12 mm
Mechanical and chemical
Videos
SDS
Documents

Documents

Product Certifications