Learn More
Ansell™ VersaTouch™ 92-200 Series Single-Use Blue Nitrile Gloves
Product Code. 12647051
Click to view available options
Size:
7
8
9
10
Improved manufacturing processes for longer wear, greater tear strength and better grip in dry, wet and fatty conditions. A thinner glove with enhanced fingertip flexibility and improved overall comfort.
  • Textured fingertips provide good comfort and tactility for sensitive, precise handling.
  • Latex-free: no risk of Type 1 allergies.
  • Approved for food contact applications.
  • Ideal for handling all fatty food.
TRUSTED_SUSTAINABILITY
Specifications
Exterior Finish Fully Textured
Material Nitrile
Size 9
Thickness (English) 2.9 mil
Thickness (Metric) 0.07 mm
Model 922009
Series 92-200