Learn More
Thermo Scientific™ Versette™ Automated Liquid Handler
Support a wide range of applications with this compact handler, featuring user-friendly programming, reliable performance and a choice of 96- or 384-channel pipetting heads.
Brand: Thermo Scientific™ 650-INSTR
Additional Details : Weight : 36.00000kg
Description
Support a wide range of applications with the Thermo Scientific™ Versette™ Automated Liquid Handler, a compact handler featuring user-friendly programming, reliable performance and a choice of 96- or 384-channel pipetting heads. An ideal choice for the liquid handling needs of most laboratories, it supports 96/384 plate replication, plate stamping and serial dilution procedures.
Support a wide range of applications, including high-throughput Mass Spectrometric Immunoassay (MSIA), plate stamping, plate reformatting, serial dilution and many other applications.
96-and 384-channel pipetting heads:
- Total volume range 0.5-300μL
- All pipetting heads include RFID tags for self-identification
Consumables for optimized performance:
- All pipetting heads utilize Thermo Scientific™ D.A.R.T.'S
Compact size:
- Six position stage uses a unique dual-level design to minimize space requirements
- Fits on standard bench
User-friendly programming options:
- Simple pipetting procedures can be created using visual on-board graphical display
- For more complex protocols and advanced editing, use Thermo Scientific™ ControlMate™ software included with purchase of the unit
- Intuitive software; after minimal training, no specialist needed for protocol writing and revisions
- Free WebEx ControlMate SW tutorial and applications support
Specifications
Versette Instrument | |
96, 384 | |
96-, 384- well plates with different heights, storage tubes in rack (Thermo Scientific 0.5 to 1.4mL) | |
Windows XP, Windows Vista (32-and 64-bit), Windows 7 (32- and 64-bit), WIN 10 | |
68 cm | |
68 cm | |
55 cm | |
54.4 kg (base system - heads/pipetting module not included), 36 kg (base - tower), 18.1 kg (6 position stage), 11.8 kg (NTC pipetting module) | |
100/240 V |
Versette instrument (includes base, pipetting module and a 6-position stage) | |
0.5 to 300 μL | |
RS-232, USB | |
27 in. | |
27 in. | |
22 in. | |
120 lb. (base system - heads/pipetting module not included), 80 lb. (base - tower), 40 lb. (6 position stage), 26 lb. (NTC pipetting module) | |
100 to 240V 50/60Hz | |
50/60 Hz |