missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ Versilon™ 2001 Multichannel Stopper Tubes
Click to view available options
Diameter (Metric) Inside:
2.79 mm
Unit Size:
Each
Description
- Always comes with an RFID-chip carrier on which important tubing parameters (inner diameter, wall thickness, service life expectancy) are saved and available for the pump’s software
- Pump is capable to start instantly in the flow rate and target volume mode through the available tubing works-factor with this RFID-chip carrier and the pump head identification
- Stopper 104
- Violet-white in color
Specifications
Specifications
| Diameter (Metric) Inside | 2.79 mm |
| Overall Length (Metric) | 381 mm |
| Size | Stopper 104 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction