Learn More
Texwipe™ Vertex™ Dry Cleanroom Wipers
Sterile dry cleanroom wipers made from 100% polyester material with a sealed edge processed on the fully automated Vertex™ manufacturing system. Vertex™ process advances cleanroom wiper manufacturing technology, bringing it to the highest level in the industry.
Brand: Texwipe™ TX3221
722.71 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Dry wipers are designed for high sorption capacity which reduces the number of wipes needed for spill control
- The sealed edge minimizes particle release from the wiper edges
- Gamma irradiated and sterile validated to a Sterility Assurance Level of 10⁻⁶ according to AAMI Guidelines
- Low levels of ions, NVRs (nonvolatile residues), particles and fibers (low linting wipes) for use in critical cleaning applications
- Designed for high sorption capacity - ideal for spill control, cleaning and solution application
- Tested for cytotoxicity and bacterial endotoxins for an extra level of contamination control assurance
- Meets USP <797> and USP <800> requirements and is autoclave safe
Specifications
Polyester | |
ISO Class 3 - 7, Class 1 - 10,000, EU Grade A - D | |
28 x 28 cm | |
5 x 20 Wipers/Bag, 5 Bags/Cs |