missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Verti-Gel Mini Tetrad PAGE System
Click to view available options
Includes:
100 x 100mm blotting module, caster and external stand, four notched/plain plates with 0.75mm spacer, four plate, four 12-well 0.75mm comb, four cassettes, eight pads
100 x 100mm blotting module, caster and external stand, four notched/plain plates with 1.5mm spacer, four plates, four 12-well 1.5mm combs, four cassettes, eight pads
100 x 100mm sliding clamps, caster and external stand, four notched/plain plates with 0.75mm bonded spacers, four notched plates, four 12-well 0.75mm combs
100 x 100mm sliding clamps, caster and external stand, four notched/plain plates with 1mm bonded spacers, four notched plates, four 12-well 1mm combs
Unit Size:
Each
Specifications
Specifications
| Type | Verti-Gel Mini Tetrad PAGE System |
| Gel Dimensions | 75 x 80 mm |
| Includes | 100 x 100mm blotting module, caster and external stand, four notched/plain plates with 0.75mm spacer, four plate, four 12-well 0.75mm comb, four cassettes, eight pads |
| Material | Injection-molded |
| No. of Gels | 4 |
| Thickness (Metric) | 0.5 mm, 0.75 mm, 1 mm, 1.5 mm, 2 mm combs |
| Volume (Metric) Buffer | 250 to 1200 mL |
| Dimensions (L x W x H) | 190 x 130 x 150 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction