missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Vertical Gel Tank

Incorporates a sealing system which is compatible with all major types of 80mm x 100mm and 100mm x 100mm precast gel. Fisherbrand™ Vertical Gel Tank's ultra soft silicone seals and pressure bars which surround the glass plates guarantee leak proof gel casting.

Brand:  Fisherbrand™ FVS20WAVESYS

 View more versions of this product

Product Code. 12623546

  • £808.00 / Each
Estimated Shipment: 13-06-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Incorporates a sealing system which is compatible with all major types of 80mm x 100mm and 100mm x 100mm precast gel. Gel casting and running utilise the same insert, no transfer of glass plates during gel casting is necessary. The insert contains sliding clamps allowing very rapid set up of both hand cast and precast gels. Ultra soft silicone seals and pressure bars which surround the glass plates guarantee leak proof gel casting. 2mm thick glass plates prevent breakage and have bonded spacers for convenience. Accessory electroblotting and tube gel modules are available which use the same outer tank and lid.
  • Injection molded construction, durable and leakproof
  • Compatible with all 80mm x 100mm and 100mm x 100mm precast gels
  • Low buffer volumes
  • Run up to four gels
  • Interchangable modules for IEF/2D electrophoresis and electroblotting in a universal tank For Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations.
Specifications

Specifications

80 mm x 100 mm and 100 mm x 100 mm PreCast Gels
20x20cm Plates, 2x1mm 24-well Combs, Cooling Coil, Casting Base
0.01 kg
Product Suggestions

Product Suggestions

Videos

Videos

  • Fisherbrand and Fisher BioReagents Focus on electrophoresis

  • Fisherbrand Meet the Family

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Vertical Gel Tank > Weight: 0.0125kg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.