Learn More
Scientific Industries SI™ Vertical Tube Holder
Requires no additional attachment for use. Scientific Industries SI™ Vertical Tube Holder snaps 15 to 50mL tubes and microtubes into place.
Brand:  Scientific Industries SI™ SI-V512
 View more versions of this product
Product Code. 11321474
  • £119.00 / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Holds tubes vertically
  • Snaps on to any Vortex-Genie 2 Mixer and the Vortex-Genie Pulse
  • Optimized for high speed vortexing a small number of tubes
  • No additional attachment necessary
Specifications
Specifications
15 mL
Vortex-Genie 2, Vortex-Genie Pulse
12