Learn More
Applied Biosystems™ VetMAX™ Xeno™ Internal Positive Control - VIC™ Assay
The VetMAX™ Xeno™ Internal Positive Control (IPC) - VIC™ Assay is a primer-probe mix that detects the Xeno internal positive control. The resultant Xeno data is used to determine the validity of diagnostic test results.
£114.00 - £506.00
Specifications
Format | Kit |
---|---|
Label or Dye | VIC |
Product Line | VetMAX™, Xeno™ |
Shipping Condition | Dry Ice |
Species | Not Species-Specific |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
30057779
|
Applied Biosystems™
A29767 |
500 Reactions |
£506.00
500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30057766
|
Applied Biosystems™
A29765 |
100 Reactions |
£114.00
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Xeno IPC assay is introduced during the qPCR preparation step and carried through the animal health PCR workflow.
Features of the VetMAX Xeno IPC - VIC Assay include:
- Provides confidence that qPCR test results are accurate and actionable
- Easily integrates into any workflow
- Greatly reduces the likelihood of false negatives
The VetMAX Xeno IPC - VIC Assay comes in a 25X concentration and easily integrates into animal health PCR workflows, regardless of the target assay, master mix, or sample preparation reagents already in place. Coupled with VetMAX™ Xeno Internal Positive Control RNA or VetMAX™ Xeno Internal Positive Control DNA, our proprietary design offers a verification layer to help ensure the qPCR test results are accurate and actionable by greatly reducing the likelihood of false negatives.
Xeno IPC assays are included in most VetMAX kits and have been successfully benchmarked against millions of genomes including those relevant to animal health. A VetMAX™ Xeno™ Internal Positive Control - LIZ™ Assay (Cat. No. A29766) is also available.
Specifications
Kit | |
VetMAX™, Xeno™ | |
Not Species-Specific |
VIC | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.