Learn More
Tocris Bioscience™ VH 032 amide-PEG2-amine
VH 032-PEG2-amine
Brand: Tocris Bioscience™ 6907/25
Additional Details : Weight : 0.00970kg
Description
VH 032 amide-PEG2-amine is a functionalized von-Hippel-Lindau (VHL) protein ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a PEG linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.This product has been recently renamed. The previous name for this product was VH 032 - linker 12
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
(2S,4R)-1-((S)-2-(2-(2-(2-Aminoethoxy)ethoxy)acetamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide | |
VH 032 - linker 12 | |
C28H41N5O6S.2HCl | |
Functionalized von-Hippel-Lindau (VHL) Ligand |
2341796-76-5 | |
25 mg | |
648.64 | |
95% |