missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Vial, Cuvette and Test Tube Holders for Orion™ AQ7100 and AQ8100 Spectrophotometers

Quickly adapt your Orion™ AquaMate 7100 & 8100 Spectrophotometer to work with multiple sample vessels

Brand:  Thermo Scientific™ AQX1LWLVH

 View more versions of this product

Product Code. 16459835

  • £31.32 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Easy to change & clean Orion™ AquaMate 7100 & 8100 Spectrophotometer sample holders are designed for maximum flexibility.

Easily adapt your Orion™ AquaMate 7100 & 8100 Spectrophotometer to work with multiple sample vessels

Magnetic retention makes these holders quick and easy to clean and change to adapt to your analytical needs.

  • AQX1RNDVH: Vial / Test Tube Holder for vials and test tube from 12 to 25 mm OD for Orion™ AquaMate 7100 & 8100 Spectrophotometers
  • AQX1SQVH: Standard 10 mm square single-cell holder for Orion™ AquaMate 7100 & 8100 Spectrophotometers
  • AQX1LWLVH: Long path rectangular cell holder for 20-100 mm pathlength cuvettes for Orion™ AquaMate 7100 & 8100 Spectrophotometers
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Long path rectangular cell holder assembled in a removable compartment liner.
Orion AquaMate 7100 & 8100 Spectrophotometers
1 Cell Holder
Rectangular
Long Path Cell Holder
20 to 100 mm path length cells
20 to 100 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers