Learn More
Fisherbrand™ Vial Decapper Pliers
Fisherbrand™ Vial Decapper Pliers are designed to easily remove aluminum crimp tops from vials and bottles.
£71.70 - £86.85
Specifications
Material Stainless Steel
Certifications/Compliance ISO 9002, cGMP
Type Plier-style Decapper
View More Specs
Products 2
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
17837658
View Documents
Fisherbrand™
13-820-043
8 mm
£71.70
Each
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
16153167
View Documents
Fisherbrand™
13820047
30 mm
£86.85
Each
Estimated Shipment: 30-07-2025
to see stock.
Add to basket
Description
Description
Fisherbrand™ Vial Decapper Pliers are designed to easily remove aluminum crimp tops from vials and bottles. Available in single- or double-action formats, all pliers are smooth and simple to operate. Durable, high-quality construction ensures consistent, dependable performance.
  • Easily remove aluminum crimp-tops from vials and bottles
  • Smooth and simple to operate
  • Durable, high-quality construction offering consistent, dependable performance
  • Padded-handles allow for maximum grip and comfort to relieve stress tension from hands
  • Made from one-piece, corrosion-resistant ASTM-standard stainless steel
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
  • Overall length: 159 mm (6.25 in.)
Certifications
ISO 9001-02, GMP, cGMP
Specifications
Specifications
Stainless Steel
ISO 9002, cGMP
Plier-style Decapper
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Vial Decapper Pliers