missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Victoria Blue B, pure

Product Code. p-2374489 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10021901

Brand: Thermo Scientific Chemicals 190190250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2580-56-5 | C33H32ClN3 | 506.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2580-56-5
Molecular Formula C33H32ClN3
Molecular Weight (g/mol) 506.09
MDL Number MFCD00011878
InChI Key LLWJPGAKXJBKKA-UHFFFAOYSA-N
Synonym Basic Blue 26, C.I. 44045
PubChem CID 17407
ChEBI CHEBI:87651
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]naphthalen-1-ylidene]-phenylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[NH+]C3=CC=CC=C3)C4=CC=CC=C24)C5=CC=C(C=C5)N(C)C.[Cl-]

Specifications

Melting Point 206.0°C
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol and ether
Infrared Spectrum Authentic
Lambda Max 599 to 605nm (in ethanol)
Formula Weight 506.09
Loss on Drying 5% max. (60°C)
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Brown
Quantity 25 g
Physical Form Glistening Crystalline Powder
Chemical Name or Material Victoria blue B
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Victoria Blue B
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.