Learn More
Ammonium Phosphate Dibasic (Colorless Crystals), Fisher BioReagents
Product Code. 10306553
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
Product Code. 10306553
Brand: Fisher BioReagents BP361500
Request Bulk or Custom Formats
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 7783-28-0 | H9N2O4P | 132.06 g/mol
Poly Bottle; 500g
Ammonium Phosphate Dibasic is suitable for use in many biological applications.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ammonium Phosphate Dibasic
CAS 7783-28-0
Color White
pH 7.9 to 8.3
Physical Form Crystalline Solid
Quantity 500 g
Assay Percent Range 98.0 to 101.0 %
Molecular Formula H9N2O4P
Linear Formula (NH4)2HPO4
MDL Number MFCD00010891
Merck Index 15, 540
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
SMILES N.N.OP(O)(O)=O
IUPAC Name phosphoric acid diamine
Molecular Weight (g/mol) 132.06
PubChem CID 24540
ChEBI CHEBI:63051
Formula Weight 132.06
Percent Purity 98.0 to 101.0%
Identification Pass Test
Packaging Poly Bottle
Density 1.619g/cm³