Learn More
Cole-Parmer™ Vinyl Color-Coded Adhesive Tape
Shop All Cole-Parmer Products
Click to view available options
Color:
Blue
Green
Orange
Red
White
Yellow
Unit Size:
Each
Description
- Tapes are on a 3" (7.6 cm) dia core
- Optional dispenser provides easy tearing of tape
- Ideal for color-coding electrical boxes, conduit or pipes
- Tough vinyl tape resists scuffs
- Adhesive sticks to most surfaces
- Order multiple roll dispenser separately
Specifications
Specifications
| Color | Orange |
| Material | Vinyl |
| Product Type | Color-coded Adhesive Tape |
| For Use With (Application) | Color-coding electrical boxes, conduit or pipes |
| Length (Metric) | 32.91 m |
| Width (Metric) | 25.4 mm |
| Diameter (Metric) | 76 mm |