Learn More
Masterflex™ NPT Male Adapter to Hose Barb
Click to view available options
Tubing Size:
1/4 in. NPT to 1/4 in. ID
3/4 in. NPT to 3/4 in. ID
Unit Size:
Pack of 10
Pack of 5
Description
- Available in a variety of materials and configurations.
- Adapts from a NPT male thread to a hose barb.
Specifications
Specifications
| Description | Barb x NPT(M) |
| Material | Polypropylene |
| Packaging Quantity | 5/Pk. |
| Tubing Size | 3/4 in. NPT to 3/4 in. ID |
| Sterilization Method | Ethylene Oxide, Gamma |
| Product Type | NPT Male Adapter to Hose Barb, 3/4 in. NPT to 3/4 in. ID |