missing translation for 'onlineSavingsMsg'
Learn More

Vilazodone-d8, MedChemExpress

Product Code. 16438627
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 16438627

Brand: Medchem Express HY14261S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vilazodone D8 is the a deuterium labeled vilazodone, which is a combined serotonin specific reuptake inhibitor (SSRI) and 5-HT1A receptor partial agonist.

Vilazodone D8 is the a deuterium labeled vilazodone, which is a combined serotonin specific reuptake inhibitor (SSRI) and 5-HT1A receptor partial agonist.

Specifications

Chemical Name or Material Vilazodone-d8
CAS 1794789-93-7
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C26H19D8N5O2
Color White
SMILES N#CC1=CC=C(NC=C2CCCCN3C([2H])([2H])C([2H])([2H])N(C4=CC=C(OC(C(N)=O)=C5)C5=C4)C([2H])([2H])C3([2H])[2H])C2=C1
Industry Type Neuroscience-Neuromodulation
Molecular Weight (g/mol) 449.57
Quantity 5 mg
Formula Weight 449.57
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.