Learn More
Vileda Professional™ CleanTech™ Duo Microfiber Laminated Mop, Sterile
For cleaning/disinfecting floors, walls and ceilings. Vileda Professional CleanTech Duo Microfiber Laminated Mop, Sterile are designed for use in cleanrooms up to class ISO 5/GMP A and B/FED209D Class 100.
Brand:  Vileda Professional™ 147544
Additional Details : Weight : 3.96000kg
Product Code. 17882496
  • £1098.20 / Pack of 45
Estimated Shipment: 30-08-2024
to see stock.
Add to basket
Description
Description
  • Microfiber laminated cleanroom foam
  • Double sided for effective cleaning and disinfecting
  • Adhesive-free lamination techniques used to prevent additional contamination to products
  • Suitable for bucket and pre-prepared method
  • Proved bacteria reduction of 99.99%
  • Validated to the 10-6 SAL per ISO 11137
  • Individually double bagged with Lot # and Indicator Dot on each package
  • Certificate of Sterility available upon request