missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Villin Monoclonal Antibody (CWWB1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512227
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-12227 targets Villin in IHC (P) applications and shows reactivity with Human samples. The MA5-12227 immunogen is human Villin protein.
Villin is part of cytoskeleton associated with the microvillar actin core bundle of gastrointestinal and renal brush border. It can interact with actin in a Ca2+ and phosphoinositide-regulated manner. Villin is a very specific marker for adenocarcinomas of the gastrointestinal tract and the pancreas. It is also expressed in some Merkel cell tumor and adenocarcinoma of the lung, prostate, ovarian and kidney.Specifications
Villin | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P09327, Q62468 | |
VIL1 | |
Human Villin protein | |
500 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
CWWB1 | |
Unconjugated | |
VIL1 | |
D2S1471; OTTHUMP00000207159; Vil; Vil1; villin 1; villin-1 | |
Mouse | |
Protein G | |
RUO | |
22349, 7429 | |
4° C | |
Liquid |