missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Vimentin

£779.00 - £1930.00

Specifications

Molecular Weight (g/mol) 57
Name Vimentin
Packaging Poly Micro Tube
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10174484
View Documents
Fisher BioReagents™
BP2545-250
250 μg
£1930.00
250µg
Please to purchase this item. Need a web account? Register with us today!
10092994
View Documents
Fisher BioReagents™
BP2545-100
100 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

57
Vimentin
Poly Micro Tube
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisher BioReagents™ Vimentin

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.