missing translation for 'onlineSavingsMsg'
Learn More

Vimentin Mouse anti-Human, Mouse, Rat, Clone: 3H9D1, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60330-1-IG-150UL

 View more versions of this product

Product Code. 16897425

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Vimentin is a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a domit, pulverulent cataract.
Specifications

Specifications

Vimentin
Monoclonal
0.8 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08670, P20152, P31000
VIM
Vimentin Fusion Protein Ag0489
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
3H9D1
Unconjugated
VIM
VIM, Vimentin
Mouse
Protein G
RUO
22352, 7431, 81818
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers