Learn More
Vinculin Monoclonal Antibody (7F9), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£141.00 - £353.00
Specifications
Antigen | Vinculin |
---|---|
Clone | 7F9 |
Concentration | 0.5 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15567676
|
Invitrogen
14-9777-80 |
25 μg |
£141.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15577676
|
Invitrogen
14-9777-82 |
100 μg |
£353.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The monoclonal antibody 7F9 (VIIF9) recognizes human, mouse, rat, and avian vinculin and its alternatively spliced isoform, metavinculin. Vinculin is a cytoskeletal protein associated with cell-cell and cell-matrix junctions. Vinculin is involved in the anchoring of F-actin to the membrane and the regulation of E-cadherin expression. Vinculin binds to talin, paxillin, and alpha-actinin. Disregulation of vinculin alters cell adhesion, migration, and growth, which promotes cancer invasion and metastasis. Applications Reported: This 7F9 antibody has been reported for use in western blotting, immunohistochemical staining of frozen tissue sections and formalin-fixed paraffin embedded tissue sections, immunocytochemistry, and microscopy. Applications Tested: This 7F9 antibody has been tested by immunocytochemistry on methanol-fixed or formaldehyde-fixed and permeabilized cells and can be used at less than or equal to 5 μg/mL. This antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded tissue using low pH antigen retrieval buffer and can be used at less than or equal to 5 μg/mL. The antibody has been tested by western blot on reduced and non-reduced cell lysates and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE.
Vinculin (VCL) is a cytoskeletal protein associated with cell-cell and cell-extracellular matrix adherens-type junctions. It functions as one of several interacting proteins involved in anchoring F-actin to the membrane. It has been shown that a sequence of molecular interactions might be involved in the transmembrane assembly of adhesion plaques. In the assembly of adhesion plaques, the beta subunit of integrin binds to talin. Talin binds to vinculin that interacts with alpha-actinin and possibly with itself. Since alpha-actinin binds to and cross-links actin filaments, vinculin represents a key element in the transmembrane linkage of the extracellular matrix to the cytoplasmic microfilament system. Mutations in the gene can result in cardiomyopathy dilated 1W or familial hypertrophic 15.Specifications
Vinculin | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
9430097D22; AA571387; AI462105; AW545629; CMD1W; CMH15; epididymis luminal protein 114; HEL114; metavinculin; meta-vinculin; metavinculin {ECO:0000250; MV; MVCL; RP11-178G16.3; UniProtKB:P18206}; unnamed protein product; Vcl; vcl {ECO:0000250; VIN; vinc; vinc 1; VINC1; Vinculin; vinculin {ECO:0000250 | |
VCL | |
Primary | |
4° C | |
VCL |
7F9 | |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P18206, P85972, Q64727 | |
22330, 305679, 7414 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.