missing translation for 'onlineSavingsMsg'
Learn More

Vinyl bromide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 428778000


 View more versions of this product

Product Code. 10553488

  • £348.74 / 800mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Vinyl bromide
593-60-2, 109-99-9
−17°C
C2H3Br
MFCD00000183
0.927
INLLPKCGLOXCIV-UHFFFAOYSA-N
bromoethene
11641
106.95
1M solution in THF
0.9270g/mL
AcroSeal™ Glass Bottle
H2C=CHBr
800 mL
vinyl bromide, bromoethylene, ethene, bromo, monobromoethylene, vinylbromid, bromure de vinyle, vinylbromide, polyvinyl bromide, ethylene, bromo, polyvinylbromide
BrC=C
106.95
CHEBI:51311
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Vinyl bromide, 1M solution in THF

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H350-May cause cancer.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H224-Extremely flammable liquid and vapour.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only