missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ C-Shape Vinyl Coated Lead Rings

Stabilize vessels and labware or immerse in water baths. Fisherbrand™ Vinyl-coated Lead Rings keep flasks and other labware weighted down without causing damage.

Brand:  Fisherbrand™ HS8881A

41.55 GBP valid until 2024-12-31
Use promo code "21757" to get your promotional price.


 View more versions of this product

Product Code. 11726725

  • £55.30 / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Vinyl-coated lead rings will not mark or scratch benchtops and labware
  • C-shape format allows some manual size variation for a customized fit
Specifications

Specifications

C-shape
1 Each
75 mm
0.277 kg
Lead
125 to 500 mL
42 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.