missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Vinyl Continuous Tape

Product Code. p-8001714
Click to view available options
Color:
Blue
Green
Orange
Red
White
Yellow
Length (Metric):
30 m
30.48 m
Width (Metric):
12.7 mm
29 mm
57 mm
101 mm
101.6 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 13274219

Brand: Brady™ 120904

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

B-7569 Vinyl Continuous Tape is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels.

  • B-7569 is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels.

Specifications

Certifications/Compliance BS5609:1986 Section 2 and section 3
Description Continuous Vinyl Tape for BBP3X/S3XXX/i3300 Printers
Adhesive Type Acrylic, Permanent
Coating None
Color Blue
Finish Gloss
Format Continuous
Material Vinyl
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Facility Identification, Floor marking, Hazard communication Identification, Pipe and valve marking, Safety Identification, Warehouse marking
For Use With (Equipment) BBP30, BBP31, BBP33, BBP35, BBP37, i3300, S3000, S3100, i5300
Includes 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels
Labels per Row 1
Min. Operating Temperature (Metric) -20°C
No. per Pack 1 Roll/Box
Quantity 1 Ea.
Software Type Brady Workstation, LabelMark, MarkWare
Temperature Range (English) -4°F to +167°F
Temperature Range (Metric) -20°C to +75°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Dimensions (LxW) 30 m x 12.7 mm
Length (English) 98 ft.
Length (Metric) 30 m
Width (English) 0.5 in.
Width (Metric) 12.7 mm
Alternate Part Number B30C-500-7569-BL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.