missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinyl decanoate, 95%, stab., Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
It is employed as a intermediates for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a intermediates for pharmaceutical.
Solubility
Soluble in methanol. Not miscible or difficult to mix with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is employed as a intermediates for pharmaceutical.
Solubility
Soluble in methanol. Not miscible or difficult to mix with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 0.886 |
Boiling Point | 119°C to 120°C (10 mmHg) |
Flash Point | 104°C (219°F) |
Linear Formula | CH3(CH2)8CO2CH=CH2 |
Refractive Index | 1.435 |
Quantity | 5 g |
Solubility Information | Soluble in methanol. Not miscible or difficult to mix with water. |
Formula Weight | 198.3 |
Percent Purity | 95% |
Chemical Name or Material | Vinyl decanoate, stabilized |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'